siRNA Details
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198605.3 | NP_941007.1 | 4 | 706 | ||
GenBank mRNA | AK089223.1 | - | 4 | 707 | ||
AK158142.1 | - | 4 | 728 | |||
AK166171.1 | - | 4 | 697 | |||
BC117500.1 | - | 4 | 698 | 40105921 | ||
BC117501.1 | - | 4 | 698 | 40105920 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108379, XM_006252074, XM_008770758, XM_006252075, BC166461.
Gene Ontology Categories:
