siRNA Details
Gene Symbol: Phf11d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199015.4 | NP_950180.3 | 3 | 283 | ||
GenBank mRNA | AK156583.1 | - | 3 | 264 | ||
BC030186.1 | - | 3 | 284 | 5067268 | ||
BC110663.1 | - | 3 | 240 | |||
BC145673.1 | - | 3 | 203 | 40130976 | ||
BG083212.2 | - | 3 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
