siRNA Details
Gene Symbol: Fam167a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177628.4 | NP_808296.2 | 3 | 1077 | ||
GenBank mRNA | AK079469.1 | - | 1 | 153 | ||
BC060626.1 | - | 1 | 190 | |||
BC065085.1 | - | 3 | 860 | 6849398 | ||
BC094561.1 | - | 3 | 987 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
