siRNA Details
Gene Symbol: Tdrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253755.1 | NP_001240684.1 | 12 | 2244 | ||
NM_172605.3 | NP_766193.3 | 12 | 2244 | |||
GenBank mRNA | AK028617.1 | - | 2 | 388 | ||
AK032892.1 | - | 12 | 2189 | |||
AK033390.1 | - | 12 | 2189 | |||
BC005670.1 | - | 12 | 2145 | 3153274 | ||
BC066144.1 | - | 12 | 2192 | 30435963 |
