siRNA Details
Gene Symbol: Dap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146057.3 | NP_666169.1 | 2, 3 | 302 | ||
GenBank mRNA | AK014991.1 | - | 2, 3 | 194 | ||
AK152216.1 | - | 2, 3 | 269 | |||
AK152884.1 | - | 2, 3 | 279 | |||
AK167579.1 | - | 2, 3 | 226 | |||
AK167688.1 | - | 2, 3 | 166 | |||
AK169162.1 | - | 2, 3 | 234 | |||
AK171259.1 | - | 2, 3 | 296 | |||
AK197429.1 | - | 1, 2 | 60 | |||
BC010828.1 | - | 2, 3 | 238 | 4220180 | ||
BC024876.1 | - | 2, 3 | 266 | 3155209 | ||
BC057669.1 | - | 2, 3 | 228 | 2655176 | ||
BY272917.1 | - | 2, 3 | 303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
