siRNA Details
Gene Symbol: Abra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175456.4 | NP_780665.1 | 2 | 1044 | ||
GenBank mRNA | AF504061.1 | - | 2 | 992 | ||
AK045186.1 | - | 873 | ||||
AK081696.1 | - | 2 | 1045 | |||
AK138943.1 | - | 743 | ||||
BC119198.1 | - | 2 | 1024 | 8733947 | ||
BC120842.1 | - | 2 | 1024 | 40129765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
