siRNA Details
Gene Symbol: Eny2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175009.3 | NP_778174.1 | 3, 4 | 275 | ||
GenBank mRNA | AF173297.1 | - | 3, 4 | 295 | ||
AK007877.1 | - | 3, 4 | 226 | |||
AK016562.1 | - | 3, 4 | 1660 | |||
AK032957.1 | - | 3, 4 | 275 | |||
BC048361.1 | - | 3, 4 | 281 | 4012930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
