siRNA Details
Gene Symbol: Nrbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144847.1 | NP_659096.1 | 11 | 911 | ||
GenBank mRNA | BC011468.1 | - | 11 | 844 | 4012880 | |
BC012437.1 | - | 11 | 911 | 3710089 | ||
BC025042.1 | - | 6 | 373 | |||
BC068117.1 | - | 16 | 1559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
