siRNA Details
Gene Symbol: Arhgap39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168288.1 | NP_001161760.1 | 3 | 741 | ||
NM_198420.2 | NP_940812.2 | 3 | 741 | |||
GenBank mRNA | AK035479.1 | - | 2 | 446 | ||
AK141623.1 | - | 3 | 701 | |||
AK173232.1 | - | 3 | 676 | |||
BC060637.1 | - | 3 | 722 | 6849400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
