siRNA Details
Gene Symbol: Zfp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145916.2 | NP_666021.2 | 5 | 450 | ||
GenBank mRNA | AK144887.1 | - | 5 | 662 | ||
AK168948.1 | - | 5 | 405 | |||
BC011501.1 | - | 5 | 468 | 3584546 | ||
BC062103.1 | - | 4 | 2052 | |||
BU613201.1 | - | 5 | 447 | |||
U29499.1 | - | 3 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
