siRNA Details
Gene Symbol: Zfp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145916.2 | NP_666021.2 | 5 | 2132 | ||
GenBank mRNA | AK144887.1 | - | 5 | 2344 | ||
AK168948.1 | - | 5 | 2087 | |||
BC011501.1 | - | 5 | 2150 | 3584546 | ||
BC062103.1 | - | 4 | 3733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
