siRNA Details
Gene Symbol: Tmem184b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253817.1 | NP_001240746.1 | 5 | 627 | ||
NM_001253819.1 | NP_001240748.1 | 5 | 648 | |||
NM_001253820.1 | NP_001240749.1 | 3 | 193 | |||
NM_172608.1 | NP_766196.1 | 5 | 673 | |||
GenBank mRNA | AK028196.1 | - | 5 | 631 | ||
AK029129.1 | - | 5 | 651 | |||
AK048138.1 | - | 5 | 627 | |||
AK149366.1 | - | 5 | 654 | |||
AK158789.1 | - | 5 | 674 | |||
AK170086.1 | - | 5 | 651 | |||
AK202130.1 | - | 1 | 27 | |||
BC046959.1 | - | 5 | 627 | 6314456 | ||
CX239857.1 | - | 3 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
