siRNA Details
Gene Symbol: Tmem184b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253817.1 | NP_001240746.1 | 8 | 1083 | ||
NM_001253819.1 | NP_001240748.1 | 8 | 1104 | |||
NM_001253820.1 | NP_001240749.1 | 6 | 649 | |||
NM_172608.1 | NP_766196.1 | 8 | 1129 | |||
GenBank mRNA | AK028196.1 | - | 8 | 1087 | ||
AK029129.1 | - | 8 | 1107 | |||
AK048138.1 | - | 8 | 1083 | |||
AK149366.1 | - | 8 | 1110 | |||
AK158789.1 | - | 8 | 1130 | |||
AK170086.1 | - | 8 | 1107 | |||
BC046959.1 | - | 8 | 1083 | 6314456 |

More Information
Other Species Targeted

Gene Ontology Categories:
