siRNA Details
Gene Symbol: Cyp2d34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145474.2 | NP_663449.2 | 6, 7 | 1042 | ||
GenBank mRNA | BC018285.1 | - | 6, 7 | 1041 | 4207630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
