siRNA Details
Gene Symbol: Ldoc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177630.3 | NP_808298.2 | 1 | 2198 | ||
GenBank mRNA | AK083881.1 | - | 1 | 2160 | ||
BC035491.1 | - | 1 | 1254 | |||
BC058638.1 | - | 1 | 1867 | 6418256 | ||
BC094559.1 | - | 1 | 2198 | 6826763 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
