siRNA Details
Gene Symbol: Zbed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181412.3 | NP_852077.1 | 3 | 2194 | ||
GenBank mRNA | AK122335.1 | - | 3 | 2164 | ||
BC052174.1 | - | 2 | 2097 | 6518433 | ||
BC092524.1 | - | 1 | 1818 |
Gene Symbol: Gm6815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133264.1 | - | 1 | 769 |
