siRNA Details
Gene Symbol: Pim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145478.2 | NP_663453.1 | 4 | 1072 | ||
GenBank mRNA | AK079623.1 | - | 4 | 795 | ||
AK149478.1 | - | 4 | 792 | |||
AK159399.1 | - | 4 | 791 | |||
AK159600.1 | - | 4 | 791 | |||
AK159670.1 | - | 4 | 768 | |||
AK168014.1 | - | 4 | 1081 | |||
AY026239.1 | - | 4 | 820 | |||
BC017621.1 | - | 4 | 1072 | 4924687 | ||
BC026639.1 | - | 4 | 761 | 4985551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
