siRNA Details
Gene Symbol: Senp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144851.5 | NP_659100.1 | 13 | 1666 | ||
GenBank mRNA | AK028565.1 | - | 13 | 1408 | ||
AK028762.1 | - | 5 | 2640 | |||
AK035723.1 | - | 14 | 1849 | |||
AK054521.1 | - | 14 | 1798 | |||
AK088597.1 | - | 13 | 1410 | |||
BC023129.1 | - | 12 | 1380 | 3595534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
