siRNA Details
Gene Symbol: Cpped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146067.3 | NP_666179.2 | 2 | 327 | ||
GenBank mRNA | AK029379.1 | - | 2 | 251 | ||
AK029442.1 | - | 2 | 350 | |||
AK032288.1 | - | 2 | 290 | |||
AK041329.1 | - | 2 | 300 | |||
AK049721.1 | - | 2 | 328 | |||
AK080876.1 | - | 2 | 285 | |||
AK133977.1 | - | 2 | 320 | |||
AK134718.1 | - | 2 | 328 | |||
BC025476.1 | - | 2 | 248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
