siRNA Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF402611.1 | - | 4 | 651 | ||
AF402612.1 | - | 3 | 548 | |||
AF402613.1 | - | 4 | 698 | |||
AK028302.1 | - | 1 | 100 | |||
AK030390.1 | - | 4 | 801 | |||
AK034504.1 | - | 3 | 641 | |||
AK043656.1 | - | 4 | 696 | |||
AK054242.1 | - | 4 | 639 | |||
AK134961.1 | - | 4 | 714 | |||
AK149626.1 | - | 4 | 702 | |||
AV137422.1 | - | 1 | 122 | |||
BB569027.2 | - | 1 | 100 | |||
BC129902.1 | - | 4 | 738 | 40111637 | ||
BC129903.1 | - | 4 | 739 | 40111639 | ||
BC145523.1 | - | 4 | 650 |
