siRNA Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF402611.1 | - | 3 | 381 | ||
AF402612.1 | - | 2 | 278 | |||
AF402613.1 | - | 3 | 428 | |||
AK030390.1 | - | 3 | 531 | |||
AK043656.1 | - | 3 | 426 | |||
AK054242.1 | - | 3 | 369 | |||
AK134961.1 | - | 3 | 444 | |||
AK149626.1 | - | 3 | 432 | |||
AK166883.1 | - | 3 | 384 | |||
BB617683.1 | - | 3 | 531 | |||
BC028644.1 | - | 3 | 303 | |||
BC129902.1 | - | 3 | 468 | 40111637 | ||
BC129903.1 | - | 3 | 469 | 40111639 | ||
BC145523.1 | - | 3 | 380 | |||
CJ120679.1 | - | 3 | 452 |
