siRNA Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF402611.1 | - | 4 | 893 | ||
AF402612.1 | - | 3 | 790 | |||
AF402613.1 | - | 4 | 940 | |||
AK028302.1 | - | 1 | 342 | |||
AK030390.1 | - | 4 | 1043 | |||
AK034504.1 | - | 3 | 883 | |||
AK043656.1 | - | 4 | 938 | |||
AK054242.1 | - | 4 | 881 | |||
AK134961.1 | - | 4 | 956 | |||
AK149626.1 | - | 4 | 944 | |||
BB569027.2 | - | 1 | 342 | |||
BC129902.1 | - | 4 | 980 | 40111637 | ||
BC129903.1 | - | 4 | 981 | 40111639 | ||
BC145523.1 | - | 4 | 892 |
