siRNA Details
Gene Symbol: Fam43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177632.3 | NP_808300.1 | 1 | 1942 | ||
GenBank mRNA | AK083036.1 | - | 1 | 1942 | ||
AK137950.1 | - | 1 | 1943 | |||
BC022623.1 | - | 1 | 535 | |||
BC026202.1 | - | 2 | 152 | |||
BC099947.1 | - | 1 | 1955 | 30088416 | ||
BC138882.1 | - | 1 | 1556 | 8861904 | ||
BC138884.1 | - | 1 | 1532 | 8861906 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
