siRNA Details
Gene Symbol: 1700021K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200038.1 | NP_001186967.1 | 20 | 2915 | ||
NM_172615.4 | NP_766203.1 | 19 | 2870 | |||
GenBank mRNA | AK030368.1 | - | 19 | 2844 | ||
AK042428.1 | - | 19 | 2808 | |||
AK122221.1 | - | 20 | 2879 | |||
AK136223.1 | - | 12 | 1552 | |||
AK170002.1 | - | 20 | 2808 | |||
AK170223.1 | - | 20 | 2766 | |||
BC037474.1 | - | 2 | 569 | |||
BC057307.1 | - | 20 | 2819 | |||
BC060601.1 | - | 20 | 2888 | 5706123 | ||
BC067390.1 | - | 19 | 2855 | 30432960 |
