siRNA Details
Gene Symbol: Zdhhc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146073.3 | NP_666185.3 | 7 | 1728 | ||
GenBank mRNA | AF542387.1 | - | 8 | 1397 | ||
AK041496.1 | - | 7 | 1411 | |||
AK046719.1 | - | 7 | 1716 | |||
AK080845.1 | - | 7 | 1732 | |||
BC059814.1 | - | 7 | 1771 | 6813525 | ||
BC086758.1 | - | 8 | 784 |
