siRNA Details
Gene Symbol: Zdhhc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146073.3 | NP_666185.3 | 6 | 1654 | ||
GenBank mRNA | AF542387.1 | - | 7 | 1323 | ||
AK041496.1 | - | 6 | 1337 | |||
AK041648.1 | - | 7 | 1650 | |||
AK046719.1 | - | 6 | 1642 | |||
AK080845.1 | - | 6 | 1658 | |||
BC059814.1 | - | 6 | 1697 | 6813525 | ||
BC086758.1 | - | 7 | 710 |
