siRNA Details
Gene Symbol: D17Wsu92e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033279.3 | NP_001028451.2 | 4 | 764 | ||
NM_001044719.2 | NP_001038184.1 | 4 | 764 | |||
GenBank mRNA | AK143016.1 | - | 4 | 676 | ||
AK161603.1 | - | 4 | 765 | |||
AK165011.1 | - | 4 | 764 | |||
BC132324.1 | - | 5 | 690 | 40130601 | ||
BC132326.1 | - | 5 | 690 | 40130603 | ||
BC137573.1 | - | 4 | 764 | 8860592 | ||
BC137576.1 | - | 4 | 764 | 8860595 | ||
BC145403.1 | - | 4 | 764 | 9053961 | ||
BU922193.1 | - | 3 | 362 | |||
CF536829.1 | - | 1 | 17 | |||
CX220817.1 | - | 4 | 539 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001039607, XM_008772799, XM_008772801, XM_008772800, BC112320.
Gene Ontology Categories:
