siRNA Details
Gene Symbol: Btbd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027060.1 | NP_081336.1 | 10 | 1765 | ||
NM_172618.2 | NP_766206.1 | 10 | 1765 | |||
GenBank mRNA | AK006275.1 | - | 5 | 651 | ||
AK029578.1 | - | 10 | 1928 | |||
AK029907.1 | - | 3 | 405 | |||
AK052664.1 | - | 10 | 1649 | |||
AK153973.1 | - | 10 | 1692 | |||
BC037031.1 | - | 8 | 1232 | |||
BC057897.1 | - | 10 | 1643 | 5031346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
