siRNA Details
Gene Symbol: Adgrf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081178.1 | NP_001074647.1 | 17 | 3918 | ||
GenBank mRNA | AK173020.1 | - | 11 | 3037 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
