siRNA Details
Gene Symbol: Prr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195673.1 | NP_001182602.1 | 3 | 866 | ||
GenBank mRNA | BC172076.1 | - | 1 | 1401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
