siRNA Details
Gene Symbol: Dlgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027712.3 | NP_081988.3 | 2 | 1106 | ||
NM_177639.6 | NP_808307.2 | 2 | 1106 | |||
GenBank mRNA | AK031410.1 | - | 6 | 1796 | ||
AK033901.1 | - | 5 | 1639 | |||
AK035763.1 | - | 5 | 1657 | |||
AK043683.1 | - | 5 | 1694 | |||
AK147448.1 | - | 6 | 1923 | |||
AK220515.1 | - | 5 | 1986 | |||
AY243846.2 | - | 1 | 941 | |||
BC062120.1 | - | 2 | 1107 | 6822047 | ||
BC094369.1 | - | 1 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
