siRNA Details
Gene Symbol: Lclat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081071.2 | NP_001074540.1 | 6 | 1264 | ||
NM_001177967.1 | NP_001171438.1 | 6 | 1243 | |||
NM_001177968.1 | NP_001171439.1 | 6 | 1232 | |||
GenBank mRNA | BC147499.1 | - | 6 | 1182 | 9056361 | |
BC150884.1 | - | 6 | 1171 | 9087795 | ||
BC158076.1 | - | 6 | 1203 | 9088168 | ||
BC158079.1 | - | 6 | 1203 | 9088171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
