siRNA Details
Gene Symbol: Wac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146298.2 | NP_001139770.1 | 2, 3 | 464 | ||
NM_001282093.1 | NP_001269022.1 | 2, 3 | 305 | |||
NM_153085.4 | NP_694725.3 | 2, 3 | 464 | |||
GenBank mRNA | AF320996.1 | - | 2, 3 | 244 | ||
AI316950.1 | - | 2, 3 | 464 | |||
AK156780.1 | - | 2, 3 | 289 | |||
AK172416.1 | - | 2, 3 | 288 | |||
AK173274.1 | - | 2, 3 | 166 | |||
BC020146.1 | - | 2, 3 | 276 | |||
BC057178.1 | - | 2, 3 | 97 | |||
BC080851.1 | - | 2, 3 | 422 | 6848738 | ||
BF461147.1 | - | 2, 3 | 132 | |||
CB249484.1 | - | 2, 3 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
