siRNA Details
Gene Symbol: Gjd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153086.5 | NP_694726.2 | 2 | 713 | ||
GenBank mRNA | AJ414562.1 | - | 2 | 522 | ||
AK034663.1 | - | 2 | 714 | |||
AK053016.1 | - | 2 | 665 | |||
BC137984.1 | - | 2 | 690 | 8861003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
