siRNA Details
Gene Symbol: Morf4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092293.1 | - | 5, 6 | 426 | 5068516 |
Gene Symbol: Gm7420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141442.1 | - | 3134 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001011999, XM_008766445, BC078910.
