siRNA Details
Gene Symbol: Ino80c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172625.2 | NP_766213.1 | 3, 4 | 389 | ||
GenBank mRNA | AK077290.1 | - | 3, 4 | 401 | ||
AK083718.1 | - | 3, 4 | 390 | |||
BC060961.1 | - | 3, 4 | 407 | 6740719 |

More Information
Other Species Targeted

Gene Ontology Categories:
