siRNA Details
Gene Symbol: Fhod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289654.1 | NP_001276583.1 | 24 | 5107 | ||
NM_001289655.1 | NP_001276584.1 | 25 | 5464 | |||
NM_175276.4 | NP_780485.2 | 26 | 5560 | |||
GenBank mRNA | AB078608.1 | - | 26 | 4738 | ||
AB078609.1 | - | 24 | 4285 | |||
AB100088.1 | - | 25 | 4642 | |||
AK220413.1 | - | 22 | 3724 | |||
BC056371.1 | - | 16 | 3190 | |||
BC060607.1 | - | 11 | 2516 | |||
BC138046.1 | - | 24 | 4207 | 8861065 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
