siRNA Details
Gene Symbol: Apbb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146085.1 | NP_666197.1 | 3 | 600 | ||
GenBank mRNA | AK141878.1 | - | 3 | 502 | ||
AK171634.1 | - | 3 | 551 | |||
BC024136.1 | - | 3 | 356 | |||
BC024809.1 | - | 3 | 600 | 5357681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
