siRNA Details
Gene Symbol: Gm94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033280.2 | NP_001028452.1 | 2, 3 | 265 | ||
GenBank mRNA | AK131907.1 | - | 2, 3 | 266 | ||
AK131908.1 | - | 2, 3 | 266 | |||
BC147098.1 | - | 2, 3 | 254 | 9056146 | ||
BC147099.1 | - | 2, 3 | 254 | 9056134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
