siRNA Details
Gene Symbol: Prdm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033281.3 | NP_001028453.1 | 6 | 1583 | ||
GenBank mRNA | AK133910.1 | - | 6 | 1584 | ||
AK135836.1 | - | 3 | 457 | |||
AK135840.1 | - | 3 | 457 | |||
BC139295.1 | - | 6 | 1613 | 8862317 | ||
DQ340564.1 | - | 6 | 1159 | |||
DQ340565.1 | - | 6 | 1078 |

More Information
Other Species Targeted

Gene Ontology Categories:
