siRNA Details
Gene Symbol: Gm4841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034859.3 | NP_001030031.2 | 2 | 1100 | ||
GenBank mRNA | AK156469.1 | - | 2 | 1106 | ||
BC150900.1 | - | 2 | 1100 | 9087811 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034859.3 | NP_001030031.2 | 2 | 1100 | ||
GenBank mRNA | AK156469.1 | - | 2 | 1106 | ||
BC150900.1 | - | 2 | 1100 | 9087811 |