siRNA Details
Gene Symbol: BC023105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC023105.1 | - | 2 | 354 | 3989161 | |
BF163606.1 | - | 2 | 354 |
Gene Symbol: Gm4841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034859.3 | NP_001030031.2 | 2 | 238 | ||
GenBank mRNA | AK156469.1 | - | 2 | 244 | ||
BC150900.1 | - | 2 | 238 | 9087811 |
