siRNA Details
Gene Symbol: Piezo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039485.4 | NP_001034574.4 | 52 | 8919 | ||
GenBank mRNA | AK031422.1 | - | 11 | 1578 | ||
AK034722.1 | - | 6 | 574 | |||
BC075677.1 | - | 13 | 2010 | |||
BC147606.1 | - | 11 | 1472 | 9056596 | ||
BC172109.1 | - | 11 | 1396 | |||
HQ215521.1 | - | 52 | 7999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
