siRNA Details
Gene Symbol: Piezo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039485.4 | NP_001034574.4 | 48, 49 | 8483 | ||
GenBank mRNA | AK031235.1 | - | 7, 8 | 2485 | ||
AK031422.1 | - | 7, 8 | 1142 | |||
AK034722.1 | - | 2, 3 | 214 | |||
BC026614.1 | - | 5, 6 | 871 | |||
BC075677.1 | - | 9, 10 | 1574 | |||
BC147606.1 | - | 7, 8 | 1036 | 9056596 | ||
BC172109.1 | - | 7, 8 | 1036 | |||
HQ215521.1 | - | 48, 49 | 7563 |

More Information
Other Species Targeted

Gene Ontology Categories:
