siRNA Details
Gene Symbol: Piezo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039485.4 | NP_001034574.4 | 48 | 8463 | ||
GenBank mRNA | AK031235.1 | - | 7 | 2465 | ||
AK031422.1 | - | 7 | 1122 | |||
AK034722.1 | - | 2 | 194 | |||
BC026614.1 | - | 5 | 851 | |||
BC075677.1 | - | 9 | 1554 | |||
BC147606.1 | - | 7 | 1016 | 9056596 | ||
BC172109.1 | - | 7 | 1016 | |||
HQ215521.1 | - | 48 | 7543 |
