siRNA Details
Gene Symbol: Alpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037294.1 | NP_001032371.1 | 5 | 4523 | ||
GenBank mRNA | AK132555.1 | - | 2 | 2851 | ||
AY044451.1 | - | 1 | 2245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
