siRNA Details
Gene Symbol: Cplx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145493.1 | NP_663468.1 | 3 | 463 | ||
GenBank mRNA | AK044262.1 | - | 3 | 471 | ||
AY264291.1 | - | 3 | 352 | |||
BC016082.1 | - | 3 | 463 | 4501240 | ||
BC029011.1 | - | 3 | 411 | 5357047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
