siRNA Details
Gene Symbol: Mppe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172630.2 | NP_766218.1 | 5, 6 | 1129 | ||
GenBank mRNA | AK041262.1 | - | 5, 6 | 1130 | ||
AK154265.1 | - | 5, 6 | 983 | |||
AK158446.1 | - | 5, 6 | 852 | |||
AK172000.1 | - | 5, 6 | 968 | |||
AK172496.1 | - | 5, 6 | 884 | |||
BC045146.1 | - | 5, 6 | 755 | 6306245 | ||
BC086456.1 | - | 5, 6 | 802 | 4949311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
