siRNA Details
Gene Symbol: Cep76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081073.1 | NP_001074542.1 | 5 | 784 | ||
GenBank mRNA | AK035879.1 | - | 4 | 564 | ||
BC119216.1 | - | 5 | 665 | 8733965 | ||
BC119218.1 | - | 5 | 665 | 8733967 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
