siRNA Details
Gene Symbol: Rnf165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164504.1 | NP_001157976.1 | 7 | 915 | ||
GenBank mRNA | AK133955.1 | - | 4 | 302 | ||
BB619563.1 | - | 4 | 302 | |||
CF745657.1 | - | 2 | 89 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: BC166566.
Gene Ontology Categories:
